Assay Details
| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013456.2 | NP_038484.1 | 13 - 14 | 1584 | 80 | |
| GenBank mRNA | AF093775.1 | - | 13 - 14 | 1559 | 80 | |
| BC111890.1 | - | 13 - 14 | 1550 | 40060598 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Actn3
Genomic Map