Assay Details
| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013456.2 | NP_038484.1 | 16 - 17 | 2041 | 107 | |
| GenBank mRNA | AF093775.1 | - | 16 - 17 | 2016 | 107 | |
| AK134757.1 | - | 1 - 2 | 145 | 107 | ||
| BC111890.1 | - | 16 - 17 | 2007 | 40060598 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Actn3
Genomic Map