| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100003978.mm.1 |
NM_013529.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003978.mm.1 |
NM_013529.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003978.mm.1 |
TC1100000526.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100003977.mm.1 |
NM_013529.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003977.mm.1 |
NM_013529.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003977.mm.1 |
TC1100000526.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100003976.mm.1 |
NM_013529.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003976.mm.1 |
NM_013529.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100003976.mm.1 |
TC1100000526.mm.1 |
MTA TC |