Assay Details
| Gene Symbol: Gfpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013529.3 | NP_038557.1 | 7 - 8 | 676 | 70 | |
| GenBank mRNA | AB016780.1 | - | 7 - 8 | 642 | 70 | |
| AK132821.1 | - | 6 - 7 | 522 | 70 | ||
| BC031928.1 | - | 7 - 8 | 644 | 4167189 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gfpt2
Genomic Map