Assay Details
| Gene Symbol: Mt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013603.2 | NP_038631.1 | 1 - 2 | 227 | 81 | |
| GenBank mRNA | AK140209.1 | - | 1 - 2 | 228 | 81 | |
| BC059725.1 | - | 1 - 2 | 125 | 6600430 | 81 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK140209.1,BC059725.1,NM_013603.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
energy reserve metabolic process
cholesterol catabolic process
zinc II ion transport
cellular metal ion homeostasis
cellular zinc ion homeostasis
response to oxidative stress
negative regulation of autophagy
positive regulation of gene expression
positive regulation of cell death
astrocyte development
regulation of response to food
leptin-mediated signaling pathway
cellular response to oxidative stress
negative regulation of apoptotic process
positive regulation of catalytic activity
negative regulation of neuron apoptotic process
cellular lipid catabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of neurogenesis
cadmium ion homeostasis
regulation of protein glycosylation
negative regulation of necrotic cell death
positive regulation of ERK1 and ERK2 cascade
positive regulation of lysosomal membrane permeability
positive regulation of oxygen metabolic process
zinc ion binding
metal ion binding
energy reserve metabolic process
cholesterol catabolic process
zinc II ion transport
cellular metal ion homeostasis
cellular zinc ion homeostasis
response to oxidative stress
negative regulation of autophagy
positive regulation of gene expression
positive regulation of cell death
astrocyte development
regulation of response to food
leptin-mediated signaling pathway
cellular response to oxidative stress
negative regulation of apoptotic process
positive regulation of catalytic activity
negative regulation of neuron apoptotic process
cellular lipid catabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of neurogenesis
cadmium ion homeostasis
regulation of protein glycosylation
negative regulation of necrotic cell death
positive regulation of ERK1 and ERK2 cascade
positive regulation of lysosomal membrane permeability
positive regulation of oxygen metabolic process
zinc ion binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P320971 | NM_013603.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P320971 | AK140209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P320971 | BC059725.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P320971 | NM_013603.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P320971 | AK140209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P320971 | BC059725.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595441 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141630 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P320971 | NM_013603.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P320971 | AK140209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P320971 | BC059725.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P320971 | NM_013603.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P320971 | AK140209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P320971 | BC059725.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26075786 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141630 | - | - |
| Whole Mouse Genome G4122A | A_52_P320971 | NM_013603.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P320971 | AK140209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P320971 | BC059725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P320971 | NM_013603.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P320971 | AK140209.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P320971 | BC059725.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120611 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138458 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141630 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07083935 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07240821 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07083947 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07272043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26075736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06785209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07083931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07348204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07083945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07193833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06897351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07103131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07197997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07083942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06781526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07434523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07135531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07064733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06966005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26075717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07103132 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10574020 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574022 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574021 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574019 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420575_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10574018 | - | - |
| Mouse Genome 430 2.0 Array | 1420575_at | - | - |
| Mouse Genome 430 2.0 Array | 1420575_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17751_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17751_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800008417.mm.1 | NM_013603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008417.mm.1 | NM_013603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008417.mm.1 | TC0800001093.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008419.mm.1 | NM_013603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008419.mm.1 | NM_013603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008419.mm.1 | TC0800001093.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008418.mm.1 | NM_013603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008418.mm.1 | NM_013603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008418.mm.1 | TC0800001093.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800008420.mm.1 | NM_013603.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008420.mm.1 | NM_013603.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800008420.mm.1 | TC0800001093.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420575_PM_at | - | - |
| Mouse Expression Set 430 | 1420575_at | - | - |
| Murine Genome U74v2 Set | 95340_PM_at | - | - |
| Murine Genome U74v2 Set | 95340_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420575_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420575_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420575_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420575_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 873028 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1450537 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001450537 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2764651 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2764651 | - | - |
| mouse-6 v1.1 (Illumina) | scl017751.1_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1450537 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl017751.1_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450537 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2764651 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2764651 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl017751.1_8-S | - | - |

Mt3
Genomic Map