Assay Details
| Gene Symbol: Usp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013700.1 | NP_038728.1 | 15 - 16 | 2001 | 68 | |
| GenBank mRNA | AK194053.1 | - | 1 - 2 | 79 | 68 | |
| BC066993.1 | - | 15 - 16 | 1979 | 5689229 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp5
Genomic Map