Assay Details
| Gene Symbol: Yme1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013771.5 | NP_038799.1 | 5 - 6 | 725 | 90 | |
| GenBank mRNA | AF090430.1 | - | 5 - 6 | 703 | 90 | |
| AK032273.1 | - | 5 - 6 | 713 | 90 | ||
| AK045141.1 | - | 5 - 6 | 710 | 90 | ||
| AK079175.1 | - | 5 - 6 | 685 | 90 | ||
| AK137665.1 | - | 5 - 6 | 677 | 90 | ||
| AY136286.1 | - | 5 - 6 | 541 | 90 | ||
| AY136287.1 | - | 5 - 6 | 694 | 90 | ||
| BC007128.1 | - | 5 - 6 | 686 | 3591123 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yme1l1
Genomic Map