Assay Details
| Gene Symbol: Abcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013806.2 | NP_038834.2 | 24 - 25 | 3502 | 67 | |
| GenBank mRNA | AF227274.1 | - | 24 - 25 | 3406 | 67 | |
| AF282772.1 | - | 24 - 25 | 3406 | 67 | ||
| AF282773.1 | - | 24 - 25 | 3406 | 67 | ||
More Information
Set Membership: |
|

Abcc2
Genomic Map