Assay Details
| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013822.5 | NP_038850.1 | 1 - 2 | 348 | 70 | |
| GenBank mRNA | AF171092.1 | - | 1 - 2 | 353 | 70 | |
| AK137092.1 | - | 1 - 2 | 248 | 70 | ||
| BC058675.1 | - | 1 - 2 | 160 | 6834418 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jag1
Genomic Map