Assay Details
| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013822.5 | NP_038850.1 | 11 - 12 | 1658 | 68 | |
| GenBank mRNA | AF171092.1 | - | 11 - 12 | 1663 | 68 | |
| AK137092.1 | - | 11 - 12 | 1558 | 68 | ||
| BC058675.1 | - | 11 - 12 | 1470 | 6834418 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jag1
Genomic Map