Assay Details
| Gene Symbol: Jag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013822.5 | NP_038850.1 | 21 - 22 | 2839 | 60 | |
| GenBank mRNA | AF171092.1 | - | 21 - 22 | 2844 | 60 | |
| AK137092.1 | - | 21 - 22 | 2739 | 60 | ||
| AK162228.1 | - | 4 - 5 | 341 | 60 | ||
| BC058675.1 | - | 21 - 22 | 2651 | 6834418 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jag1
Genomic Map