Assay Details
| Gene Symbol: Gtse1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168672.1 | NP_001162143.1 | 10 - 11 | 2080 | 102 | |
| NM_013882.2 | NP_038910.1 | 9 - 10 | 2050 | 102 | ||
| GenBank mRNA | AJ222580.1 | - | 9 - 10 | 2020 | 102 | |
| AK081582.1 | - | 10 - 11 | 2043 | 102 | ||
| BC027213.1 | - | 9 - 10 | 2051 | 4039841 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtse1
Genomic Map