| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600015070.mm.1 |
TC0600001926.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015070.mm.1 |
NM_013930.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015070.mm.1 |
NM_013930.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015069.mm.1 |
TC0600001926.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015069.mm.1 |
NM_013930.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015069.mm.1 |
NM_013930.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015071.mm.1 |
TC0600001926.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015071.mm.1 |
NM_013930.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015071.mm.1 |
NM_013930.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015067.mm.1 |
TC0600001926.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015067.mm.1 |
NM_013930.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015067.mm.1 |
NM_013930.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015072.mm.1 |
TC0600001926.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015072.mm.1 |
NM_013930.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015072.mm.1 |
NM_013930.4 |
Refseq |