Assay Details
| Gene Symbol: Il27ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016671.3 | NP_057880.2 | 5 - 6 | 712 | 57 | |
| GenBank mRNA | AF053005.1 | - | 5 - 6 | 680 | 57 | |
| AF265243.1 | - | 5 - 6 | 691 | 57 | ||
| AK040457.1 | - | 3 - 4 | 818 | 57 | ||
| AK133753.1 | - | 5 - 6 | 727 | 57 | ||
| BC032878.1 | - | 5 - 6 | 712 | 1244998 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il27ra
Genomic Map