Assay Details
| Gene Symbol: Il27ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016671.3 | NP_057880.2 | 7 - 8 | 982 | 66 | |
| GenBank mRNA | AF053005.1 | - | 7 - 8 | 950 | 66 | |
| AF265243.1 | - | 7 - 8 | 961 | 66 | ||
| AK133753.1 | - | 7 - 8 | 997 | 66 | ||
| BC032878.1 | - | 7 - 8 | 982 | 1244998 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il27ra
Genomic Map