Assay Details
| Gene Symbol: Uba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016682.2 | NP_057891.1 | 3 - 4 | 309 | 138 | |
| GenBank mRNA | AK146925.1 | - | 3 - 4 | 355 | 138 | |
| AK151765.1 | - | 3 - 4 | 314 | 138 | ||
| AK152415.1 | - | 3 - 4 | 333 | 138 | ||
| AK152977.1 | - | 3 - 4 | 314 | 138 | ||
| AK163451.1 | - | 3 - 4 | 305 | 138 | ||
| AK164826.1 | - | 3 - 4 | 325 | 138 | ||
| AK165321.1 | - | 3 - 4 | 331 | 138 | ||
| AK166133.1 | - | 3 - 4 | 341 | 138 | ||
| AK168673.1 | - | 3 - 4 | 321 | 138 | ||
| AK169168.1 | - | 3 - 4 | 305 | 138 | ||
| AK184258.1 | - | 3 - 4 | 217 | 138 | ||
| AK192304.1 | - | 3 - 4 | 217 | 138 | ||
| AK194855.1 | - | 3 - 4 | 217 | 138 | ||
| AK204958.1 | - | 3 - 4 | 217 | 138 | ||
| AK219985.1 | - | 3 - 4 | 217 | 138 | ||
| BC054768.1 | - | 3 - 4 | 335 | 6415071 | 138 | |
| U35833.1 | - | 3 - 4 | 310 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uba2
Genomic Map