Assay Details
| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016696.4 | NP_057905.1 | 2 - 3 | 565 | 78 | |
| GenBank mRNA | AF185613.1 | - | 2 - 3 | 568 | 78 | |
| AK154899.1 | - | 2 - 3 | 608 | 78 | ||
| BC062902.1 | - | 2 - 3 | 394 | 6810413 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpc1
Genomic Map