Assay Details
| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016696.4 | NP_057905.1 | 5 - 6 | 1256 | 103 | |
| GenBank mRNA | AF185613.1 | - | 5 - 6 | 1259 | 103 | |
| AK031768.1 | - | 224 | 103 | |||
| AK154584.1 | - | 3 - 4 | 390 | 103 | ||
| AK154899.1 | - | 5 - 6 | 1299 | 103 | ||
| BC039232.1 | - | 1 - 2 | 63 | 103 | ||
| BC062902.1 | - | 5 - 6 | 1085 | 6810413 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpc1
Genomic Map