Assay Details
| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016696.4 | NP_057905.1 | 6 - 7 | 1378 | 80 | |
| GenBank mRNA | AF185613.1 | - | 6 - 7 | 1381 | 80 | |
| AK031768.1 | - | 346 | 80 | |||
| AK154584.1 | - | 4 - 5 | 512 | 80 | ||
| AK154899.1 | - | 6 - 7 | 1421 | 80 | ||
| AK206330.1 | - | 1 - 2 | 69 | 80 | ||
| BC039232.1 | - | 2 - 3 | 185 | 80 | ||
| BC062902.1 | - | 6 - 7 | 1207 | 6810413 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpc1
Genomic Map