Assay Details
| Gene Symbol: Grb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016719.1 | NP_057928.1 | 6 - 7 | 930 | 121 | |
| GenBank mRNA | AF155647.1 | - | 6 - 7 | 930 | 121 | |
| AK010849.1 | - | 6 - 7 | 947 | 121 | ||
| AK010903.1 | - | 6 - 7 | 947 | 121 | ||
| AK034134.1 | - | 1 - 2 | 1221 | 121 | ||
| AK147041.1 | - | 6 - 7 | 939 | 121 | ||
| BC138581.1 | - | 6 - 7 | 836 | 8861601 | 121 | |
| BC145913.1 | - | 6 - 7 | 836 | 40131216 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grb14
Genomic Map