Assay Details
| Gene Symbol: Usp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016808.2 | NP_058088.2 | 4 - 5 | 1135 | 90 | |
| NM_198091.2 | NP_932759.1 | 3 - 4 | 591 | 90 | ||
| NM_198092.2 | NP_932760.2 | 4 - 5 | 1232 | 90 | ||
| GenBank mRNA | AB041799.1 | - | 3 - 4 | 552 | 90 | |
| AF079565.1 | - | 3 - 4 | 193 | 90 | ||
| AK138799.1 | - | 5 - 6 | 1433 | 90 | ||
| AY255637.1 | - | 3 - 4 | 560 | 90 | ||
| AY255638.1 | - | 4 - 5 | 1114 | 90 | ||
| AY255639.1 | - | 4 - 5 | 1068 | 90 | ||
| AY255640.1 | - | 4 - 5 | 1113 | 90 | ||
| BC017517.1 | - | 4 - 5 | 1114 | 4506362 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp2
Genomic Map