Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900003266.mm.1 |
NM_016808.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003266.mm.1 |
NM_016808.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003266.mm.1 |
NM_198091.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003266.mm.1 |
NM_198091.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003266.mm.1 |
NM_198092.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003266.mm.1 |
NM_198092.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003266.mm.1 |
TC0900000497.mm.1 |
MTA TC |