Assay Details
| Gene Symbol: Dnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110831.1 | NP_001104301.1 | 4 - 5 | 598 | 53 | |
| NM_016878.4 | NP_058574.3 | 4 - 5 | 422 | 53 | ||
| GenBank mRNA | AF005051.1 | - | 3 - 4 | 532 | 53 | |
| AK038646.1 | - | 598 | 53 | |||
| AK052001.1 | - | 400 | 53 | |||
| AK160084.1 | - | 4 - 5 | 359 | 53 | ||
| AK182360.1 | - | 1 - 2 | 81 | 53 | ||
| BC092232.1 | - | 4 - 5 | 350 | 30551654 | 53 | |
More Information
Set Membership: |
|

Dnpep
Genomic Map