Assay Details
Gene Symbol: Ptprcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | D10105.1 | - | 1 - 2 | 40 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the D10105.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AK090093.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | U03856.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AI627045.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AK165203.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AW049152.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AK090093.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | U03856.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AI627045.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AK165203.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P247359 | AW049152.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493190 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132036 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146989 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127526 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AK090093.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | U03856.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AI627045.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AK165203.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AW049152.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AK090093.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | U03856.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AI627045.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AK165203.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P247359 | AW049152.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AK090093.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | U03856.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AI627045.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AK165203.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AW049152.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AK090093.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | U03856.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AI627045.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AK165203.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P247359 | AW049152.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921416 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921434 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921436 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921415 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921427 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921435 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921425 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921432 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921433 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921430 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921419 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921421 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31921424 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146989 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127526 | - | - |
Whole Mouse Genome G4122A | A_51_P247359 | AK090093.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P247359 | U03856.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P247359 | AI627045.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P247359 | AK165203.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P247359 | AW049152.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P247359 | AK090093.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P247359 | U03856.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P247359 | AI627045.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P247359 | AK165203.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P247359 | AW049152.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138799 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127526 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146989 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14222824 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14222828 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14371126 | - | - |
Agilent Mouse cDNA Microarray (G4104A) | 642720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31921416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14371126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14463643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921427 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921424 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14222822 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921436 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921433 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921434 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31921417 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10460375 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10460373 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10460372 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10460374 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448511_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10460371 | - | - |
Mouse Genome 430 2.0 Array | 1448511_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1448511_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19265_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19265_PM_at | - | - |
Affymetrix Murine Genome 6500A Array | Msa13780_at | - | - |
Mouse Expression Set 430 | 1448511_PM_at | - | - |
Mouse Expression Set 430 | 1448511_at | - | - |
Murine Genome U74v2 Set | 99478_at | - | - |
Murine Genome U74v2 Set | 99478_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448511_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1448511_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448511_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448511_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6650075 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6650075 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006650075 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2652719 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2652719 | - | - |
mouse-6 v1.1 (Illumina) | scl52802.3.52_13-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6650075 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl52802.3.52_13-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650075 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2652719 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2652719 | - | - |
