Assay Details
| Gene Symbol: Oat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016978.2 | NP_058674.1 | 2 - 3 | 342 | 59 | |
| GenBank mRNA | AK128914.1 | - | 2 - 3 | 333 | 59 | |
| AK145877.1 | - | 2 - 3 | 343 | 59 | ||
| AK146413.1 | - | 2 - 3 | 287 | 59 | ||
| AK168851.1 | - | 2 - 3 | 291 | 59 | ||
| AK185243.1 | - | 1 - 2 | 126 | 59 | ||
| AK185430.1 | - | 1 - 2 | 126 | 59 | ||
| BC008119.1 | - | 2 - 3 | 275 | 3498240 | 59 | |
| X64837.1 | - | 1 - 2 | 316 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oat
Genomic Map