Assay Details
| Gene Symbol: Icos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017480.2 | NP_059508.2 | 3 - 4 | 553 | 113 | |
| GenBank mRNA | AB023132.1 | - | 3 - 4 | 537 | 113 | |
| AF216748.1 | - | 3 - 4 | 508 | 113 | ||
| AF257230.1 | - | 3 - 4 | 538 | 113 | ||
| AJ250559.1 | - | 3 - 4 | 553 | 113 | ||
| AK030827.1 | - | 3 - 4 | 546 | 113 | ||
| BC034852.1 | - | 3 - 4 | 540 | 1364228 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Icos
Genomic Map