Assay Details
| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018743.4 | NP_061213.2 | 8 - 9 | 1876 | 78 | |
| GenBank mRNA | AF015811.1 | - | 5 - 6 | 552 | 78 | |
| AF406611.1 | - | 7 - 8 | 1387 | 78 | ||
| AK045235.1 | - | 8 - 9 | 1770 | 78 | ||
| AK077561.1 | - | 8 - 9 | 1760 | 78 | ||
| AK083589.1 | - | 8 - 9 | 1770 | 78 | ||
| AK161270.1 | - | 8 - 9 | 1739 | 78 | ||
| AK162033.1 | - | 5 - 6 | 681 | 78 | ||
| AK169257.1 | - | 8 - 9 | 1770 | 78 | ||
| AK199339.1 | - | 2 - 3 | 165 | 78 | ||
| AK216721.1 | - | 2 - 3 | 212 | 78 | ||
| AK218609.1 | - | 2 - 3 | 192 | 78 | ||
| AY489184.1 | - | 7 - 8 | 945 | 78 | ||
| BC031767.1 | - | 7 - 8 | 1230 | 4457846 | 78 | |
| BC057860.1 | - | 5 - 6 | 550 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agpat6
Genomic Map