Assay Details
| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018743.4 | NP_061213.2 | 10 - 11 | 2012 | 69 | |
| GenBank mRNA | AF015811.1 | - | 7 - 8 | 688 | 69 | |
| AF406611.1 | - | 9 - 10 | 1523 | 69 | ||
| AK045235.1 | - | 10 - 11 | 1906 | 69 | ||
| AK077561.1 | - | 10 - 11 | 1896 | 69 | ||
| AK083589.1 | - | 10 - 11 | 1906 | 69 | ||
| AK161270.1 | - | 10 - 11 | 1875 | 69 | ||
| AK162033.1 | - | 7 - 8 | 817 | 69 | ||
| AK167876.1 | - | 10 - 11 | 1921 | 69 | ||
| AK169257.1 | - | 10 - 11 | 1906 | 69 | ||
| AK184759.1 | - | 4 - 5 | 348 | 69 | ||
| AK203007.1 | - | 4 - 5 | 287 | 69 | ||
| AK216721.1 | - | 4 - 5 | 348 | 69 | ||
| AY489184.1 | - | 9 - 10 | 1081 | 69 | ||
| BC031767.1 | - | 9 - 10 | 1366 | 4457846 | 69 | |
| BC057860.1 | - | 7 - 8 | 686 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agpat6
Genomic Map