Assay Details
| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018743.4 | NP_061213.2 | 11 - 12 | 2143 | 64 | |
| GenBank mRNA | AF015811.1 | - | 8 - 9 | 819 | 64 | |
| AF406611.1 | - | 10 - 11 | 1654 | 64 | ||
| AK045235.1 | - | 11 - 12 | 2037 | 64 | ||
| AK077561.1 | - | 11 - 12 | 2027 | 64 | ||
| AK083589.1 | - | 11 - 12 | 2037 | 64 | ||
| AK161270.1 | - | 11 - 12 | 2006 | 64 | ||
| AK162033.1 | - | 8 - 9 | 948 | 64 | ||
| AK167876.1 | - | 11 - 12 | 2052 | 64 | ||
| AK169257.1 | - | 11 - 12 | 2037 | 64 | ||
| BC031767.1 | - | 10 - 11 | 1497 | 4457846 | 64 | |
| BC057860.1 | - | 8 - 9 | 817 | 64 | ||
More Information
Set Membership: |
|

Agpat6
Genomic Map