Assay Details
| Gene Symbol: Itih4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159299.2 | NP_001152771.1 | 10 - 11 | 1404 | 70 | |
| NM_001289632.1 | NP_001276561.1 | 10 - 11 | 1404 | 70 | ||
| NM_001289633.1 | NP_001276562.1 | 10 - 11 | 1404 | 70 | ||
| NM_018746.4 | NP_061216.2 | 10 - 11 | 1404 | 70 | ||
| GenBank mRNA | AF023919.1 | - | 10 - 11 | 1367 | 70 | |
| AK004862.1 | - | 10 - 11 | 1379 | 70 | ||
| AK004893.1 | - | 10 - 11 | 1379 | 70 | ||
| AK050016.1 | - | 10 - 11 | 1382 | 70 | ||
| AK050270.1 | - | 10 - 11 | 1379 | 70 | ||
| BC016500.1 | - | 10 - 11 | 1367 | 4211735 | 70 | |
| BC021459.1 | - | 10 - 11 | 1369 | 70 | ||
| BC092258.1 | - | 10 - 11 | 1356 | 5125112 | 70 | |
| BC094457.1 | - | 10 - 11 | 1373 | 5136326 | 70 | |
More Information
Set Membership: |
|

Itih4
Genomic Map