Assay Details
Gene Symbol: Gp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018762.1 | NP_061232.1 | 1 - 2 | 45 | 52 | |
GenBank mRNA | AB007464.1 | - | 1 - 2 | 45 | 52 | |
BC111106.1 | - | 1 - 2 | 40 | 40055578 | 52 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB007464.1,BC111106.1,NM_018762.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P153995 | NM_018762.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P153995 | AB007464.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P153995 | NM_018762.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P153995 | AB007464.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128462 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P153995 | NM_018762.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P153995 | AB007464.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P153995 | NM_018762.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P153995 | AB007464.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P153995 | NM_018762.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P153995 | AB007464.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P153995 | NM_018762.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P153995 | AB007464.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618097 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618092 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618078 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618058 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618064 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618075 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618076 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618062 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618079 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618088 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618085 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618094 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618100 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618060 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618068 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618072 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618070 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618099 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618081 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618071 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618069 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618082 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618066 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618073 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618080 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618077 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618087 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24618061 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128462 | - | - |
Whole Mouse Genome G4122A | A_51_P153995 | NM_018762.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P153995 | AB007464.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P153995 | NM_018762.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P153995 | AB007464.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100978 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128462 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05507966 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05507962 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05507961 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05507965 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05507973 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05507958 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05507959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24618111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05507959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05065107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05217460 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618077 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05274777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05217458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05431144 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05687016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618114 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05065111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618099 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05507962 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24618052 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10539821 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10539819 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10539820 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419720_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10539818 | - | - |
Mouse Genome 430 2.0 Array | 1419720_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419720_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54368_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 54368_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600007945.mm.1 | TC0600001016.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600007945.mm.1 | NM_018762.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600007945.mm.1 | NM_018762.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600007947.mm.1 | TC0600001016.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600007947.mm.1 | NM_018762.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600007947.mm.1 | NM_018762.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600007946.mm.1 | TC0600001016.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600007946.mm.1 | NM_018762.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600007946.mm.1 | NM_018762.1 | Refseq |
Mouse Expression Set 430 | 1419720_at | - | - |
Mouse Expression Set 430 | 1419720_PM_at | - | - |
Murine Genome U74v2 Set | 102311_at | - | - |
Murine Genome U74v2 Set | 102311_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419720_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419720_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419720_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419720_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 839237 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1190181 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1190181 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001190181 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248875 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1248875 | - | - |
mouse-6 v1.1 (Illumina) | scl29782.3.1_1-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1190181 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl29782.3.1_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1190181 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2842601 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2842601 | - | - |
