Assay Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018765.3 | NP_061235.2 | 6 - 7 | 832 | 69 | |
| GenBank mRNA | AF071184.1 | - | 6 - 7 | 684 | 69 | |
| AK135215.1 | - | 6 - 7 | 832 | 69 | ||
| AK159522.1 | - | 6 - 7 | 1084 | 69 | ||
| AK160559.1 | - | 6 - 7 | 569 | 69 | ||
| AK161496.1 | - | 6 - 7 | 526 | 69 | ||
| AK162009.1 | - | 6 - 7 | 574 | 69 | ||
| U40746.1 | - | 2 - 3 | 112 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wbp4
Genomic Map