Assay Details
| Gene Symbol: Abcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018795.2 | NP_061265.2 | 8 - 9 | 1000 | 64 | |
| GenBank mRNA | AB028737.1 | - | 8 - 9 | 1006 | 64 | |
| BC040400.1 | - | 8 - 9 | 1008 | 64 | ||
| EF109740.1 | - | 8 - 9 | 1000 | 64 | ||
| EF109741.1 | - | 8 - 9 | 1000 | 64 | ||
More Information
Set Membership: |
|

Abcc6
Genomic Map