Assay Details
| Gene Symbol: Abcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018795.2 | NP_061265.2 | 16 - 17 | 2069 | 60 | |
| GenBank mRNA | AB028737.1 | - | 16 - 17 | 2075 | 60 | |
| BC040400.1 | - | 16 - 17 | 2077 | 60 | ||
More Information
Set Membership: |
|

Abcc6
Genomic Map