Assay Details
| Gene Symbol: Impa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018864.5 | NP_061352.2 | 6 - 7 | 576 | 75 | |
| GenBank mRNA | AF042730.1 | - | 6 - 7 | 521 | 75 | |
| AK011770.1 | - | 6 - 7 | 514 | 75 | ||
| AK078406.1 | - | 6 - 7 | 525 | 75 | ||
| AK083238.1 | - | 6 - 7 | 510 | 75 | ||
| AK145472.1 | - | 513 | 75 | |||
| AK147778.1 | - | 6 - 7 | 536 | 75 | ||
| AK152774.1 | - | 6 - 7 | 527 | 75 | ||
| AK153457.1 | - | 6 - 7 | 543 | 75 | ||
| AK160203.1 | - | 6 - 7 | 514 | 75 | ||
| AK160467.1 | - | 6 - 7 | 508 | 75 | ||
| AK165977.1 | - | 6 - 7 | 518 | 75 | ||
| AK168457.1 | - | 6 - 7 | 551 | 75 | ||
| AK189530.1 | - | 1 - 2 | 60 | 75 | ||
| BC049080.1 | - | 6 - 7 | 517 | 6419835 | 75 | |
| BC055722.1 | - | 5 - 6 | 471 | 6413389 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Impa1
Genomic Map