Assay Details
| Gene Symbol: Lsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136071.2 | NP_001129543.1 | 2 - 3 | 298 | 68 | |
| NM_001271508.1 | NP_001258437.1 | 2 - 3 | 665 | 68 | ||
| NM_001271509.1 | NP_001258438.1 | 2 - 3 | 298 | 68 | ||
| NM_001271510.1 | NP_001258439.1 | 2 - 3 | 298 | 68 | ||
| NM_001271523.1 | NP_001258452.1 | 2 - 3 | 497 | 68 | ||
| NM_019391.3 | NP_062264.1 | 2 - 3 | 665 | 68 | ||
| GenBank mRNA | AK003081.1 | - | 2 - 3 | 247 | 68 | |
| AK031587.1 | - | 2 - 3 | 638 | 68 | ||
| AK075783.1 | - | 2 - 3 | 498 | 68 | ||
| BC003796.1 | - | 2 - 3 | 357 | 3488528 | 68 | |
| BC044736.1 | - | 2 - 3 | 253 | 68 | ||
| D49691.1 | - | 2 - 3 | 302 | 68 | ||
| M89956.1 | - | 2 - 3 | 240 | 68 | ||
| M90316.1 | - | 2 - 3 | 664 | 68 | ||
| S74179.1 | - | 2 - 3 | 211 | 68 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271523.1 transcripts.
Set Membership: |
|

Lsp1
Genomic Map