Assay Details
| Gene Symbol: Ptbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019550.2 | NP_062423.1 | 9 - 10 | 1224 | 75 | |
| GenBank mRNA | AF095718.2 | - | 9 - 10 | 1094 | 75 | |
| AK033964.1 | - | 2 - 3 | 1631 | 75 | ||
| AK135738.1 | - | 9 - 10 | 1227 | 75 | ||
| AK137348.1 | - | 9 - 10 | 1103 | 75 | ||
| BC010255.1 | - | 9 - 10 | 1110 | 3709255 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptbp2
Genomic Map