Assay Details
| Gene Symbol: Ptbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019550.2 | NP_062423.1 | 10 - 11 | 1260 | 58 | |
| GenBank mRNA | AF095718.2 | - | 10 - 11 | 1130 | 58 | |
| AK033964.1 | - | 3 - 4 | 1667 | 58 | ||
| AK135738.1 | - | 10 - 11 | 1263 | 58 | ||
| AK137348.1 | - | 10 - 11 | 1139 | 58 | ||
| BC010255.1 | - | 10 - 11 | 1146 | 3709255 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptbp2
Genomic Map