Assay Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019670.1 | NP_062644.1 | 11 - 12 | 1181 | 73 | |
| GenBank mRNA | AF094519.1 | - | 11 - 12 | 1181 | 73 | |
| AK137073.1 | - | 10 - 11 | 1270 | 73 | ||
| AK138694.1 | - | 11 - 12 | 1211 | 73 | ||
| AK161737.1 | - | 10 - 11 | 1199 | 73 | ||
| AK166882.1 | - | 11 - 12 | 1237 | 73 | ||
| AK220493.1 | - | 11 - 12 | 1245 | 73 | ||
| BC003250.1 | - | 10 - 11 | 1215 | 73 | ||
| BC028920.1 | - | 4 - 5 | 482 | 4008236 | 73 | |
| BC085191.1 | - | 11 - 12 | 1291 | 30693873 | 73 | |
| BC086779.1 | - | 11 - 12 | 1256 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap3
Genomic Map