Assay Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019670.1 | NP_062644.1 | 15 - 16 | 1583 | 112 | |
| GenBank mRNA | AF094519.1 | - | 15 - 16 | 1583 | 112 | |
| AK137073.1 | - | 14 - 15 | 1672 | 112 | ||
| AK138694.1 | - | 15 - 16 | 1613 | 112 | ||
| AK161737.1 | - | 14 - 15 | 1601 | 112 | ||
| AK166882.1 | - | 15 - 16 | 1639 | 112 | ||
| BC028920.1 | - | 8 - 9 | 884 | 4008236 | 112 | |
| BC085191.1 | - | 15 - 16 | 1693 | 30693873 | 112 | |
| BC086779.1 | - | 15 - 16 | 1658 | 112 | ||
More Information
Set Membership: |
|

Diap3
Genomic Map