Assay Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019670.1 | NP_062644.1 | 24 - 25 | 2960 | 106 | |
| GenBank mRNA | AF094519.1 | - | 24 - 25 | 2960 | 106 | |
| AK166882.1 | - | 24 - 25 | 3016 | 106 | ||
| AK220493.1 | - | 24 - 25 | 3024 | 106 | ||
| BC028920.1 | - | 17 - 18 | 2261 | 4008236 | 106 | |
| BC085191.1 | - | 24 - 25 | 3070 | 30693873 | 106 | |
| BC086779.1 | - | 24 - 25 | 3035 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap3
Genomic Map