Assay Details
| Gene Symbol: Tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252443.1 | NP_001239372.1 | 2 - 3 | 489 | 109 | |
| NM_019678.3 | NP_062652.1 | 2 - 3 | 489 | 109 | ||
| GenBank mRNA | AK008549.1 | - | 2 - 3 | 265 | 109 | |
| AK088864.1 | - | 2 - 3 | 307 | 109 | ||
| AK158539.1 | - | 2 - 3 | 355 | 109 | ||
| AK167475.1 | - | 2 - 3 | 353 | 109 | ||
| AK186338.1 | - | 1 - 2 | 109 | |||
| BC024638.1 | - | 2 - 3 | 267 | 4015300 | 109 | |
| U94662.1 | - | 2 - 3 | 391 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfg
Genomic Map