Assay Details
| Gene Symbol: Tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019678.3 | NP_062652.1 | 4 - 5 | 728 | 65 | |
| GenBank mRNA | AK008549.1 | - | 4 - 5 | 504 | 65 | |
| AK158539.1 | - | 4 - 5 | 594 | 65 | ||
| AK167475.1 | - | 4 - 5 | 592 | 65 | ||
| BC024638.1 | - | 4 - 5 | 506 | 4015300 | 65 | |
| U94662.1 | - | 4 - 5 | 630 | 65 | ||
More Information
Set Membership: |
|

Tfg
Genomic Map