Assay Details
| Gene Symbol: Rbck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083921.1 | NP_001077390.1 | 5 - 6 | 944 | 82 | |
| NM_019705.3 | NP_062679.2 | 6 - 7 | 785 | 82 | ||
| GenBank mRNA | AF124663.1 | - | 4 - 5 | 554 | 82 | |
| AK085559.1 | - | 5 - 6 | 933 | 82 | ||
| AK088591.1 | - | 5 - 6 | 906 | 82 | ||
| AK146176.1 | - | 6 - 7 | 959 | 82 | ||
| AK166075.1 | - | 5 - 6 | 912 | 82 | ||
| BC034555.1 | - | 6 - 7 | 781 | 4192310 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbck1
Genomic Map