| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000007198.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007198.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007198.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007198.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007198.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007199.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007199.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007199.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007199.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007199.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007204.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007204.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007204.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007204.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007204.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007197.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007197.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007197.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007197.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007197.mm.1 |
TC1000000904.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000007201.mm.1 |
NM_001252401.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007201.mm.1 |
NM_001252401.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007201.mm.1 |
NM_019725.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007201.mm.1 |
NM_019725.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000007201.mm.1 |
TC1000000904.mm.1 |
MTA TC |