Assay Details
| Gene Symbol: P3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042411.1 | NP_001035874.1 | 9 - 10 | 1164 | 63 | |
| NM_001286148.1 | NP_001273077.1 | 9 - 10 | 1551 | 63 | ||
| NM_019782.3 | NP_062756.2 | 9 - 10 | 1530 | 63 | ||
| NM_019783.2 | NP_062757.2 | 9 - 10 | 1530 | 63 | ||
| GenBank mRNA | AF165163.1 | - | 9 - 10 | 1496 | 63 | |
| AF165164.1 | - | 9 - 10 | 1496 | 63 | ||
| AK010578.1 | - | 9 - 10 | 1532 | 63 | ||
| AK030436.1 | - | 9 - 10 | 1528 | 63 | ||
| AK132262.1 | - | 9 - 10 | 1527 | 63 | ||
| AK159505.1 | - | 9 - 10 | 1165 | 63 | ||
| BC024047.1 | - | 9 - 10 | 1496 | 5148677 | 63 | |
| CX563255.1 | - | 5 - 6 | 422 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P3h1
Genomic Map