Assay Details
| Gene Symbol: Aatf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019816.1 | NP_062790.1 | 6 - 7 | 969 | 92 | |
| GenBank mRNA | AF222801.1 | - | 6 - 7 | 969 | 92 | |
| AK016820.1 | - | 2 - 3 | 206 | 92 | ||
| AK077789.1 | - | 6 - 7 | 993 | 92 | ||
| AK141334.1 | - | 6 - 7 | 967 | 92 | ||
| BC025080.1 | - | 6 - 7 | 956 | 2648117 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aatf
Genomic Map