Assay Details
| Gene Symbol: Ovol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019935.3 | NP_064319.1 | 2 - 3 | 594 | 80 | |
| GenBank mRNA | AF134804.1 | - | 2 - 3 | 610 | 80 | |
| AF134805.1 | - | 2 - 3 | 576 | 80 | ||
| AK003801.1 | - | 2 - 3 | 629 | 80 | ||
| AK028577.1 | - | 2 - 3 | 595 | 80 | ||
| AK132478.1 | - | 2 - 3 | 595 | 80 | ||
| AK132505.1 | - | 2 - 3 | 633 | 80 | ||
| AK132506.1 | - | 2 - 3 | 629 | 80 | ||
| BC021411.1 | - | 2 - 3 | 674 | 5006712 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ovol1
Genomic Map