Assay Details
| Gene Symbol: Dnase2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019957.4 | NP_064341.3 | 5 - 6 | 865 | 72 | |
| GenBank mRNA | AF128888.1 | - | 4 - 5 | 744 | 72 | |
| AK079224.1 | - | 5 - 6 | 866 | 72 | ||
| BC075661.1 | - | 4 - 5 | 620 | 30606058 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnase2b
Genomic Map