Assay Details
| Gene Symbol: Olfr544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020289.2 | NP_064685.2 | 2 | 932 | 73 | |
| GenBank mRNA | BC106830.2 | - | 1 | 40044631 | 73 | |
| BC126870.1 | - | 1 | 40044632 | 73 | ||
| BC126871.1 | - | 1 | 40044633 | 73 | ||
| Gene Symbol: Olfr545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146840.1 | NP_667051.1 | 1 | 73 | ||
| GenBank mRNA | BC106987.1 | - | 1 | 40043277 | 73 | |
| BC128265.1 | - | 1 | 40043266 | 73 | ||
| BC128266.1 | - | 1 | 40043268 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134387 | NM_020289.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134387 | BC106830.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134387 | BC126870.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134387 | BC126871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134387 | NM_020289.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134387 | BC106830.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134387 | BC126870.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134387 | BC126871.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P235448 | NM_020289.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P235448 | AF121982.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P235448 | BC106830.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P235448 | BC126870.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P235448 | BC126871.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25379044 | - | - |
| Whole Mouse Genome G4122A | A_51_P235448 | NM_020289.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P235448 | AF121982.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P235448 | BC106830.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P235448 | BC126870.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P235448 | BC126871.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06445591 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06566777 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06566780 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06379490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25378972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06403730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06445591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06642818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06642810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06642813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06566779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06189379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06642820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25379075 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566139 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566141 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422354_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566140 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566138 | - | - |
| Mouse Genome 430 2.0 Array | 1422354_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422354_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 257926_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 257926_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700032628.mm.1 | NM_020289.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032628.mm.1 | NM_020289.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032628.mm.1 | TC0700003860.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032630.mm.1 | NM_020289.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032630.mm.1 | NM_020289.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032630.mm.1 | TC0700003860.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700032629.mm.1 | NM_020289.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032629.mm.1 | NM_020289.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032629.mm.1 | TC0700003860.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422354_at | - | - |
| Mouse Expression Set 430 | 1422354_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422354_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422354_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422354_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422354_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 818145 | - | - |
| ABI Mouse Genome Survey Microarray | 899254 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2120102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002120102 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220645 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220645 | - | - |
| mouse-6 v1.1 (Illumina) | scl30941.1.393_192-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2120102 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30941.1.393_192-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120102 | - | - |

Olfr544
Genomic Map